1,2,3-Trichloro-4-iodobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

53814-1G 53814-500MG 53814-5G
76891-502EA 500.75 USD
76891-502 76891-504 76891-506
1,2,3-Trichloro-4-iodobenzene ≥97% research grade
1,2,3-Trichloro-4-iodobenzene
Formula: C₆H₂Cl₃I
Storage Temperature: Ambient
CAS Number: 62720-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR