1-Chloro-3-iodo-2-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

53476-25G 53476-5G
76891-462EA 315.09 USD
76891-462 76891-464
1-Chloro-3-iodo-2-methoxybenzene ≥95%
1-Chloro-3-iodo-2-methoxybenzene
Formula: C₇H₆ClIO
Storage Temperature: Ambient
MDL Number: MFCD18393315
CAS Number: 860585-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR