1-Bromo-3-fluoro-4-iodo-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53220-1G 53220-25G 53220-500MG 53220-5G
76891-416EA 230.22 USD
76891-416 76891-418 76891-420 76891-422
1-Bromo-3-fluoro-4-iodo-2-methoxybenzene ≥97%
1-Bromo-3-fluoro-4-iodo-2-methoxybenzene
Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34178678
CAS Number: 2383590-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR