2-(Benzyloxy)-1-bromo-4-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

53151-25G 53151-5G
76891-392EA 466.8 USD
76891-392 76891-394
2-(Benzyloxy)-1-bromo-4-chlorobenzene ≥97%
2-(Benzyloxy)-1-bromo-4-chlorobenzene
Formula: C₁₃H₁₀BrClO
MW: 297.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30295981
CAS Number: 690261-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR