(3,4-Dichloro-2-Fluorophenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

53135-1G 53135-25G 53135-5G 53135-500MG
76891-378EA 109.28 USD
76891-378 76891-380 76891-382 77647-564
(3,4-Dichloro-2-Fluorophenyl)(Methyl)Sulfane ≥97%
(3,4-Dichloro-2-Fluorophenyl)(Methyl)Sulfane
Formula: C₇H₅Cl₂FS
MW: 211.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786344
CAS Number: 1804421-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR