1-(Benzyloxy)-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

53064-1G 53064-25G 53064-5G
76891-338EA 94.43 USD
76891-338 76891-340 76891-342
1-(Benzyloxy)-2-iodobenzene ≥97%
1-(Benzyloxy)-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁IO
MW: 310.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14648490
CAS Number: 142523-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR