4’-Bromo-3’-methylacetophenone ≥97%
Supplier: AOBChem USA
|
Warning
|
52992-1G
52992-25G
52992-5G
76891-324EA
79.57
USD
76891-324
76891-326
76891-328
4’-Bromo-3’-methylacetophenone ≥97%
4’-Bromo-3’-methylacetophenone
Precaución: for Research Use Only
Formula:
C₉H₉BrO Storage Temperature: Refrigerator |
MDL Number:
MFCD00045017 CAS Number: 37074-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...