(4-Chloro-2-methoxy-5-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

52825-1G 52825-25G 52825-500MG 52825-5G
76891-310EA 334.19 USD
76891-310 76891-312 76891-314 76891-316
(4-Chloro-2-methoxy-5-methylphenyl)boronic acid ≥97% research grade
(4-Chloro-2-methoxy-5-methylphenyl)boronic acid
Formula: C₈H₁₀BClO₃
Storage Temperature: Ambient
CAS Number: 1393536-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR