(5-Methoxy-2,4-dimethylphenyl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

52622-1G 52622-500MG 52622-5G
76891-290EA 1003.62 USD
76891-290 76891-292 76891-294
(5-Methoxy-2,4-dimethylphenyl)boronic acid ≥95% research grade
(5-Methoxy-2,4-dimethylphenyl)boronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD28795052
CAS Number: 1421934-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR