(2-Chloro-5-ethylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

52524-1G 52524-25G 52524-500MG 52524-5G
76891-248EA 292.81 USD
76891-248 76891-250 76891-252 76891-254
(2-Chloro-5-ethylphenyl)boronic acid ≥95%
(2-Chloro-5-ethylphenyl)boronic acid
Formula: C₈H₁₀BClO₂
Storage Temperature: Ambient
MDL Number: MFCD28167189
CAS Number: 1418023-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR