1-Bromo-4-chloro-2-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52500-1G 52500-500MG 52500-5G
76891-234EA 526.21 USD
76891-234 76891-236 76891-238
1-Bromo-4-chloro-2-methoxy-3-methylbenzene ≥97%
1-Bromo-4-chloro-2-methoxy-3-methylbenzene
Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414768
CAS Number: 55289-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR