2,3-Dibromo-4-methylthiophene ≥97%

Supplier: AOBChem USA

Warning

52408-1G 52408-25G 52408-500MG 52408-5G
76891-220EA 153.84 USD
76891-220 76891-222 76891-224 76891-226
2,3-Dibromo-4-methylthiophene ≥97%
2,3-Dibromo-4-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₄Br₂S
Storage Temperature: Ambient
CAS Number: 125257-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR