2,4-Dibromo-6-fluoro-3-hydroxybenzoic acid 95% research grade

Supplier: AOBChem USA

Warning

52224-1G 52224-25G 52224-5G 52224-500MG
76891-198EA 62.6 USD
76891-198 76891-200 76891-202 77647-540
2,4-Dibromo-6-fluoro-3-hydroxybenzoic acid 95% research grade
2,4-Dibromo-6-fluoro-3-hydroxybenzoic acid
Formula: C₇H₃Br₂FO₃
Storage Temperature: Ambient
MDL Number: MFCD34178620
CAS Number: 91659-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR