2,4-Dibromo-3-chloroaniline ≥97%

Supplier: AOBChem USA

Warning

51985-1G 51985-25G 51985-5G 51985-500MG
76891-178EA 88.06 USD
76891-178 76891-180 76891-182 77647-526
2,4-Dibromo-3-chloroaniline ≥97%
2,4-Dibromo-3-chloroaniline
Formula: C₆H₄Br₂ClN
Storage Temperature: Ambient
MDL Number: MFCD22480892
CAS Number: 80026-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR