1-(5-Chloro-2-fluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

51962-25G 51962-5G
76891-154EA 453.01 USD
76891-154 76891-156
1-(5-Chloro-2-fluorophenyl)ethanol ≥97%
1-(5-Chloro-2-fluorophenyl)ethanol
Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD09907953
CAS Number: 958653-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR