1,3-Dibromo-5-tert-butyl-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Dibromo-5-(tert-butyl)-2-methoxybenzene

51675-25G 51675-5G
76891-134EA 375.56 USD
76891-134 76891-136
1,3-Dibromo-5-tert-butyl-2-methoxybenzene ≥97%
1,3-Dibromo-5-tert-butyl-2-methoxybenzene
Formula: C₁₁H₁₄Br₂O
MW: 322.0363 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233136
CAS Number: 132268-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR