2-(Benzyloxy)-1-bromo-3,5-bis(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

51505-1G 51505-500MG 51505-5G
76891-118EA 587.74 USD
76891-118 76891-120 76891-122
2-(Benzyloxy)-1-bromo-3,5-bis(trifluoromethyl)benzene ≥95%
2-(Benzyloxy)-1-bromo-3,5-bis(trifluoromethyl)benzene
Formula: C₁₅H₉BrF₆O
MW: 399.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902948
CAS Number: 2586126-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR