1-Bromo-2-(cyclopropylmethoxy)benzene ≥95% research grade

Supplier: AOBChem USA

Warning

51496-1G 51496-25G 51496-5G
76891-104EA 62.6 USD
76891-104 76891-106 76891-108
1-Bromo-2-(cyclopropylmethoxy)benzene ≥95% research grade
1-Bromo-2-(cyclopropylmethoxy)benzene
Formula: C₁₀H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD12138831
CAS Number: 494773-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR