(2,3-Difluoro-6-(methylthio)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

51473-1G 51473-25G 51473-500MG 51473-5G
76891-088EA 439.22 USD
76891-088 76891-090 76891-092 76891-094
(2,3-Difluoro-6-(methylthio)phenyl)boronic acid ≥97%
(2,3-Difluoro-6-(methylthio)phenyl)boronic acid
Formula: C₇H₇BF₂O₂S
MW: 204.002 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902946
CAS Number: 2614261-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR