2-(Difluoromethoxy)-1-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

51447-1G 51447-25G 51447-500MG 51447-5G
76891-072EA 441.34 USD
76891-072 76891-074 76891-076 76891-078
2-(Difluoromethoxy)-1-fluoro-4-iodobenzene ≥97%
2-(Difluoromethoxy)-1-fluoro-4-iodobenzene
Formula: C₇H₄F₃IO
MW: 288.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396814
CAS Number: 1261572-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR