(2-Fluoro-5-Iodo-1,3-Phenylene)Bis(Methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

51439-1G 51439-25G 51439-500MG 51439-5G
76891-056EA 292.81 USD
76891-056 76891-058 76891-060 76891-062
(2-Fluoro-5-Iodo-1,3-Phenylene)Bis(Methylsulfane) ≥97%
(2-Fluoro-5-Iodo-1,3-Phenylene)Bis(Methylsulfane)
Formula: C₈H₈FIS₂
MW: 314.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902936
CAS Number: 2586127-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR