1-Bromo-4-chloro-2-(difluoromethoxy)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

51340-1G 51340-25G 51340-500MG 51340-5G
76891-026EA 361.77 USD
76891-026 76891-028 76891-030 76891-032
1-Bromo-4-chloro-2-(difluoromethoxy)benzene ≥97% research grade
1-Bromo-4-chloro-2-(difluoromethoxy)benzene
Formula: C₇H₄BrClF₂O
MW: 257.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698391
CAS Number: 1056942-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR