(4-Bromo-2-propoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51239-1G 51239-25G 51239-500MG 51239-5G
76891-006EA 177.18 USD
76891-006 76891-008 76891-010 76891-012
(4-Bromo-2-propoxyphenyl)methanol ≥97%
(4-Bromo-2-propoxyphenyl)methanol
Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11619327
CAS Number: 1094510-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR