1-Bromo-2-(difluoromethoxy)-4-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51192-1G 51192-500MG 51192-5G
76890-994EA 558.04 USD
76890-994 76890-996 76890-998
1-Bromo-2-(difluoromethoxy)-4-fluoro-5-methylbenzene ≥97%
1-Bromo-2-(difluoromethoxy)-4-fluoro-5-methylbenzene
Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902917
CAS Number: 2167304-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR