1-Chloro-5-fluoro-4-iodo-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

50756-1G 50756-25G 50756-500MG 50756-5G
76890-966EA 144.29 USD
76890-966 76890-968 76890-970 76890-972
1-Chloro-5-fluoro-4-iodo-2-methylbenzene ≥95%
1-Chloro-5-fluoro-4-iodo-2-methylbenzene
Formula: C₇H₅ClFI
Storage Temperature: Refrigerator
MDL Number: MFCD16658720
CAS Number: 1126424-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR