(3-(Difluoromethoxy)-5-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

50605-1G 50605-25G 50605-5G 50605-500MG
76890-952EA 102.91 USD
76890-952 76890-954 76890-956 77651-438
(3-(Difluoromethoxy)-5-fluorophenyl)boronic acid ≥97%
(3-(Difluoromethoxy)-5-fluorophenyl)boronic acid
Formula: C₇H₆BF₃O₃
MW: 205.927 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18395052
CAS Number: 2055539-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR