6-Bromo-3-fluoro-o-xylene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-fluoro-2,3-dimethylbenzene, 1-Bromo-2,3-dimethyl-4-fluorobenzene

50200-25G 50200-5G
76890-932EA 286.45 USD
76890-932 76890-934
6-Bromo-3-fluoro-o-xylene ≥97% research grade
6-Bromo-3-fluoro-o-xylene
Formula: C₈H₈BrF
MW: 203.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789115
CAS Number: 52548-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR