1-Iodo-3-methoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50116-1G 50116-25G 50116-5G 50116-500MG
76890-918EA 117.76 USD
76890-918 76890-920 76890-922 77651-434
1-Iodo-3-methoxy-5-(trifluoromethyl)benzene ≥97%
1-Iodo-3-methoxy-5-(trifluoromethyl)benzene
Formula: C₈H₆F₃IO
MW: 302.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18398117
CAS Number: 868167-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR