2,4-Dibromo-3,5-dichloroaniline ≥95%

Supplier: AOBChem USA

Warning

50032-1G 50032-25G 50032-500MG 50032-5G
76890-902EA 155.96 USD
76890-902 76890-904 76890-906 76890-908
2,4-Dibromo-3,5-dichloroaniline ≥95%
2,4-Dibromo-3,5-dichloroaniline

Precaución: for Research Use Only

Formula: C₆H₃Br₂Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD09878197
CAS Number: 1000574-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR