1-Bromo-4-chloro-5-fluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49980-1G 49980-500MG 49980-5G
76890-888EA 525.15 USD
76890-888 76890-890 76890-892
1-Bromo-4-chloro-5-fluoro-2-methoxybenzene ≥97%
1-Bromo-4-chloro-5-fluoro-2-methoxybenzene
Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30297359
CAS Number: 2090323-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR