2,4-Dichloro-3-methyl benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Dichloro-3-methylbenzoic acid

49939-1G 49939-25G 49939-5G
76890-874EA 62.6 USD
76890-874 76890-876 76890-878
2,4-Dichloro-3-methyl benzoic acid ≥97% research grade
2,4-Dichloro-3-methyl benzoic acid
Formula: C₈H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16659660
CAS Number: 83277-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR