2-Chloro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-3-methylphenyl)methanol

49932-1G 49932-25G 49932-500MG 49932-5G
76890-860EA 137.92 USD
76890-860 76890-862 76890-864 76890-866
2-Chloro-3-methylbenzyl alcohol ≥97%
2-Chloro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031743
CAS Number: 61563-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR