1-Bromo-2,4-diiodobenzene ≥95%

Supplier: AOBChem USA

Warning

49571-1G 49571-25G 49571-500MG 49571-5G
76890-772EA 194.15 USD
76890-772 76890-774 76890-776 76890-778
1-Bromo-2,4-diiodobenzene ≥95%
1-Bromo-2,4-diiodobenzene
Formula: C₆H₃BrI₂
Storage Temperature: Ambient
CAS Number: 126063-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR