2,3-Difluoro-4-(methoxymethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49345-1G 49345-25G 49345-500MG 49345-5G
76890-724EA 265.23 USD
76890-724 76890-726 76890-728 76890-730
2,3-Difluoro-4-(methoxymethoxy)benzaldehyde ≥97%
2,3-Difluoro-4-(methoxymethoxy)benzaldehyde
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32902822
CAS Number: 2586126-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR