2,4-Dibromo-6-fluorotoluene ≥95%

Supplier: AOBChem USA

Warning

49204-1G 49204-25G 49204-500MG 49204-5G
76890-712EA 173.99 USD
76890-712 76890-714 76890-716 76890-718
2,4-Dibromo-6-fluorotoluene ≥95%
2,4-Dibromo-6-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
Storage Temperature: Refrigerator
MDL Number: MFCD09800775
CAS Number: 85233-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR