4-Bromo-2-fluoro-5-nitrotoluene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-5-fluoro-4-methyl-2-nitrobenzene

49199-25G 49199-5G
76890-706EA 240.83 USD
76890-706 76890-708
4-Bromo-2-fluoro-5-nitrotoluene ≥97% research grade
4-Bromo-2-fluoro-5-nitrotoluene
Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD11042845
CAS Number: 1345471-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR