1-Bromo-5-chloro-4-methyl-2-nitrobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
49195-1G
49195-25G
49195-500MG
49195-5G
76890-690EA
260.99
USD
76890-690
76890-692
76890-694
76890-696
1-Bromo-5-chloro-4-methyl-2-nitrobenzene ≥97%
1-Bromo-5-chloro-4-methyl-2-nitrobenzene
Formula:
C₇H₅BrClNO₂ MW: 250.477 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16652298 CAS Number: 1228671-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...