2,4-Dichloro-5-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

49119-1G 49119-25G 49119-500MG 49119-5G
76890-668EA 227.04 USD
76890-668 76890-670 76890-672 76890-674
2,4-Dichloro-5-methylbenzonitrile ≥97%
2,4-Dichloro-5-methylbenzonitrile
Formula: C₈H₅Cl₂N
MW: 186.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16817654
CAS Number: 33373-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR