2,3,6-Trichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48679-25G 48679-5G
76890-580EA 275.84 USD
76890-580 76890-582
2,3,6-Trichlorobenzoic acid ≥97%
2,3,6-Trichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00053310
CAS Number: 50-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR