2,3,6-Trichlorotoluene ≥97%

Supplier: AOBChem USA

Warning

48676-25G 48676-5G
76890-572EA 502.87 USD
76890-572 76890-574
2,3,6-Trichlorotoluene ≥97%
2,3,6-Trichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013623
CAS Number: 2077-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR