1,5-Dibromo-2,4-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

48656-1G 48656-25G 48656-500MG 48656-5G
76890-556EA 263.11 USD
76890-556 76890-558 76890-560 76890-562
1,5-Dibromo-2,4-dichlorobenzene ≥97%
1,5-Dibromo-2,4-dichlorobenzene
Formula: C₆H₂Br₂Cl₂
MW: 304.794 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28783264
CAS Number: 100191-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR