1,2-Difluoro-3-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

48603-25G 48603-5G
76890-546EA 275.84 USD
76890-546 76890-548
1,2-Difluoro-3-iodobenzene ≥95%
1,2-Difluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂I
MW: 239.9893 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458090
CAS Number: 64248-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR