2,3-Difluoro-4-formylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

48586-1G 48586-500MG 48586-5G
76890-534EA 944.21 USD
76890-534 76890-536 76890-538
2,3-Difluoro-4-formylbenzoic acid ≥95%
2,3-Difluoro-4-formylbenzoic acid
Formula: C₈H₄F₂O₃
MW: 186.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32639736
CAS Number: 1897016-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR