3-Bromo-5-chloro-4-iodobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3-chloro-2-iodo-5-(trifluoromethyl)benzene

48476-1G 48476-25G 48476-500MG 48476-5G
76890-518EA 423.3 USD
76890-518 76890-520 76890-522 76890-524
3-Bromo-5-chloro-4-iodobenzotrifluoride ≥97%
3-Bromo-5-chloro-4-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂BrClF₃I
Storage Temperature: Refrigerator
MDL Number: MFCD00042178
CAS Number: 175205-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR