2,4-Dibromo-6-tert-butylphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Dibromo-6-(tert-butyl)phenol

48445-1G 48445-25G 48445-5G
76890-506EA 117.76 USD
76890-506 76890-508 76890-510
2,4-Dibromo-6-tert-butylphenol ≥97%
2,4-Dibromo-6-tert-butylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂O
MW: 308.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463268
CAS Number: 15460-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR