1,4-Dibromo-2,3-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

48294-1G 48294-25G 48294-5G
76890-448EA 64.72 USD
76890-448 76890-450 76890-452
1,4-Dibromo-2,3-dichlorobenzene ≥97%
1,4-Dibromo-2,3-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂Cl₂
MW: 304.794 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878188
CAS Number: 100191-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR