1,5-Dibromo-2,3-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

48291-1G 48291-25G 48291-5G
76890-438EA 80.63 USD
76890-438 76890-440 76890-442
1,5-Dibromo-2,3-dichlorobenzene ≥97%
1,5-Dibromo-2,3-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂Cl₂
MW: 304.794 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070783
CAS Number: 81067-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR