2,2,2-trifluoro-1-(4-fluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

48184-1G 48184-25G 48184-500MG 48184-5G
76890-422EA 153.84 USD
76890-422 76890-424 76890-426 76890-428
2,2,2-trifluoro-1-(4-fluorophenyl)ethanol ≥97%
2,2,2-trifluoro-1-(4-fluorophenyl)ethanol
Formula: C₈H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD00615801
CAS Number: 50562-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR