4-Bromo-2-methoxy-1-methylsulfanylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-2-methoxyphenyl)(methyl)sulphane

48081-1G 48081-25G 48081-5G
76890-402EA 76.39 USD
76890-402 76890-404 76890-406
4-Bromo-2-methoxy-1-methylsulfanylbenzene ≥97%
4-Bromo-2-methoxy-1-methylsulfanylbenzene

Precaución: for Research Use Only

Formula: C₈H₉BrOS
MW: 233.125 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27944804
CAS Number: 144369-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR