5-Bromo-2,4-difluoroiodobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2,4-difluoro-5-iodobenzene

48007-25G 48007-5G
76890-392EA 310.85 USD
76890-392 76890-394
5-Bromo-2,4-difluoroiodobenzene ≥97% research grade
5-Bromo-2,4-difluoroiodobenzene
Formula: C₆H₂BrF₂I
MW: 318.885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458121
CAS Number: 914636-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR